Assay Details
Target Gene Details
Entrez Gene ID: | 3575 |
Gene Name: | interleukin 7 receptor |
Gene Aliases: |
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA |
Location: |
Chr.5:35856875-35879603 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL7R | NM_002185.3 | 5 | 697 | NP_002176.2 |
| NR_120485.1 | ||||
| XM_005248299.3 | 5 | 682 | XP_005248356.1 | |
| AK301220.1 | 5 | 681 | ||
| AK315251.1 | 5 | 596 | ||
| BC020717.1 | 5 | 683 | ||
| BC067537.1 | 5 | 674 | AAH67537.1 | |
| BC067538.1 | 5 | 674 | AAH67538.1 | |
| BC067539.1 | 5 | 674 | AAH67539.1 | |
| BC067540.1 | 5 | 674 | AAH67540.1 | |
| BC069999.1 | 5 | 674 | AAH69999.1 | |
| FJ529208.1 | ||||
| M29696.1 | 5 | 616 | AAA59157.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv483069 | Chr.5:35785417 - 35955982 on Build GRCh38 | Gain |
|
| esv3604707 | Chr.5:35513484 - 36501574 on Build GRCh38 | Gain |
|
| nsv830261 | Chr.5:35862982 - 36017946 on Build GRCh38 | Gain |
|
| nsv471011 | Chr.5:35860966 - 36003115 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map