Assay Details
Target Gene Details
Entrez Gene ID: | 4113 |
Gene Name: | MAGE family member B2 |
Gene Aliases: |
CT3.2, DAM6, MAGE-XP-2 |
Location: |
Chr.X:30215558-30220089 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAGEB2 | NM_002364.4 | 2 | 1159 | NP_002355.2 |
| XM_011545512.1 | 2 | 1199 | XP_011543814.1 | |
| AF015766.1 | 2 | 1157 | AAD01565.1 | |
| AK223081.1 | 2 | 1141 | BAD96801.1 | |
| BC026071.2 | 2 | 1154 | AAH26071.1 |
More Information
Set Membership: |
|

Genomic Map