Assay Details
Target Gene Details
Entrez Gene ID: | 121130 |
Gene Name: | olfactory receptor family 10 subfamily P member 1 |
Gene Aliases: |
OR10P1P, OR10P2P, OR10P3P, OR12-7, OST701 |
Location: |
Chr.12:55636892-55637833 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR10P1 | NM_206899.1 | 1 | 695 | NP_996782.1 |
| BC136914.1 | 1 | 727 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77750365] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map