Assay Details
Target Gene Details
Entrez Gene ID: | 54676 |
Gene Name: | GTP binding protein 2 |
Gene Aliases: |
- |
Location: |
Chr.6:43620481-43629246 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTPBP2 | NM_001286216.1 | 8 | 1073 | NP_001273145.1 |
| NM_019096.4 | 8 | 1268 | NP_061969.3 | |
| XM_017010976.1 | 8 | 1262 | XP_016866465.1 | |
| AB024574.1 | 9 | 1111 | BAB12431.1 | |
| AF168990.1 | 8 | 1065 | AAF78884.1 | |
| AJ420518.1 | 3 | 431 | ||
| AK000430.1 | 4 | 220 | BAA91160.1 | |
| AK290267.1 | 8 | 1073 | ||
| AL834331.1 | 8 | 1050 | CAD38999.1 | |
| BC020980.2 | 4 | 684 | AAH20980.2 | |
| BC028347.2 | 6 | 976 | AAH28347.2 | |
| BC064968.1 | 8 | 1221 | AAH64968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv5281 | Chr.6:43611090 - 43655905 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map