Assay Details
Target Gene Details
Entrez Gene ID: | 3913 |
Gene Name: | laminin subunit beta 2 |
Gene Aliases: |
LAMS, NPHS5 |
Location: |
Chr.3:49121114-49133166 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LAMB2 | NM_002292.3 | 25 | 4229 | NP_002283.3 |
| XM_005265127.3 | 26 | 4028 | XP_005265184.1 | |
| AK094050.1 | 6 | 848 | BAC04272.1 | |
| S77512.1 | 25 | 4082 | AAB34682.2 | |
| X72760.1 | 2 | 384 | CAA51288.1 | |
| X79683.1 | 26 | 4095 | CAA56130.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834686 | Chr.3:49068614 - 49233123 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map