Assay Details
Target Gene Details
Entrez Gene ID: | 6853 |
Gene Name: | synapsin I |
Gene Aliases: |
SYN1a, SYN1b, SYNI |
Location: |
Chr.X:47571901-47619857 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYN1 | NM_006950.3 | 13 | 2740 | NP_008881.2 |
| NM_133499.2 | 13 | 2702 | NP_598006.1 | |
| AL833961.1 | 5 | 1525 |
More Information
Set Membership: |
|

Genomic Map