Assay Details
Target Gene Details
Entrez Gene ID: | 8742 |
Gene Name: | tumor necrosis factor superfamily member 12 |
Gene Aliases: |
APO3L, DR3LG, TNLG4A, TWEAK |
Location: |
Chr.17:7549058-7557890 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF12 | NM_003809.2 | 7 | 647 | NP_003800.1 |
| NR_037146.1 | 8 | 886 | ||
| AF030099.1 | 7 | 568 | AAC51923.1 | |
| AF055872.1 | 7 | 608 | AAC39724.1 | |
| AY358870.1 | 7 | 608 | AAQ89229.1 | |
| BC104420.1 | 7 | 583 | AAI04421.1 |
Target Gene Details
Entrez Gene ID: | 407977 |
Gene Name: | TNFSF12-TNFSF13 readthrough |
Gene Aliases: |
TWE-PRIL |
Location: |
Chr.17:7549058-7561608 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TNFSF12-TNFSF13 | NM_172089.3 | NP_742086.1 | ||
| AY081051.1 | AAL90443.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422288 | Chr.17:6789933 - 7646709 on Build GRCh38 | Deletion |
|
| nsv574322 | Chr.17:7010333 - 7689462 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map