Assay Details
Target Gene Details
Entrez Gene ID: | 201176 |
Gene Name: | Rho GTPase activating protein 27 |
Gene Aliases: |
CAMGAP1, PP905, SH3D20, SH3P20 |
Location: |
Chr.17:45393902-45432916 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ARHGAP27 | NM_001282290.1 | 17 | 3554 | NP_001269219.1 |
| NM_199282.2 | 17 | 2962 | NP_954976.1 | |
| XM_005257128.3 | 16 | 3011 | XP_005257185.1 | |
| XM_006721745.2 | 20 | 3936 | XP_006721808.1 | |
| XM_006721747.2 | 19 | 3870 | XP_006721810.1 | |
| XM_011524468.1 | 21 | 4035 | XP_011522770.1 | |
| XM_011524469.1 | 20 | 3969 | XP_011522771.1 | |
| XM_011524470.2 | 21 | 4025 | XP_011522772.1 | |
| XM_011524471.1 | 21 | 3864 | XP_011522773.1 | |
| XM_011524472.2 | 18 | 3176 | XP_011522774.1 | |
| XM_011524474.1 | 18 | 3046 | XP_011522776.1 | |
| XM_017024311.1 | 19 | 3969 | XP_016879800.1 | |
| XM_017024312.1 | 17 | 3075 | XP_016879801.1 | |
| AK125535.1 | 17 | 2947 | BAC86196.1 | |
| BC029634.1 | 1 | 617 | AAH29634.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428342 | Chr.17:45366188 - 45573718 on Build GRCh38 | Gain |
|
| esv2758691 | Chr.17:45380520 - 45738657 on Build GRCh38 | Gain+Loss |
|
| nsv1070831 | Chr.17:45307533 - 45449134 on Build GRCh38 | Deletion |
|
| dgv1472n106 | Chr.17:45307534 - 45500034 on Build GRCh38 | Deletion |
|
| dgv123e55 | Chr.17:45393394 - 45589540 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map