Assay Details
Target Gene Details
Entrez Gene ID: | 7812 |
Gene Name: | cold shock domain containing E1 |
Gene Aliases: |
D1S155E, UNR |
Location: |
Chr.1:114716913-114758050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CSDE1 | NM_001007553.2 | NP_001007554.1 | ||
| NM_001130523.2 | NP_001123995.1 | |||
| NM_001242891.1 | NP_001229820.1 | |||
| NM_001242892.1 | NP_001229821.1 | |||
| NM_001242893.1 | NP_001229822.1 | |||
| NM_007158.5 | NP_009089.4 | |||
| AB020692.1 | ||||
| AF070542.1 | AAC28634.1 | |||
| AF077054.1 | AAD27787.1 | |||
| AK022516.1 | BAB14073.1 | |||
| AK022528.1 | BAB14080.1 | |||
| AK290146.1 | ||||
| AY049788.1 | AAL15445.1 | |||
| BC032446.1 | AAH32446.1 | |||
| BX648561.1 | ||||
| BX648780.1 | ||||
| CR749378.1 | CAH18231.1 | |||
| CR749431.1 | CAH18269.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28734 | Chr.1:114376708 - 115005298 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map