Assay Details
Target Gene Details
Entrez Gene ID: | 9908 |
Gene Name: | G3BP stress granule assembly factor 2 |
Gene Aliases: |
- |
Location: |
Chr.4:75642769-75724525 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| G3BP2 | NM_012297.4 | 12 | 1576 | NP_036429.2 |
| NM_203504.2 | 11 | 1612 | NP_987100.1 | |
| NM_203505.2 | 12 | 1711 | NP_987101.1 | |
| XM_005263382.2 | 12 | 1685 | XP_005263439.1 | |
| XM_005263383.3 | 13 | 1650 | XP_005263440.1 | |
| XM_011532441.2 | 12 | 1571 | XP_011530743.1 | |
| XM_017008876.1 | 12 | 1559 | XP_016864365.1 | |
| XM_017008877.1 | 12 | 1738 | XP_016864366.1 | |
| XM_017008878.1 | 11 | 1460 | XP_016864367.1 | |
| XM_017008879.1 | 11 | 1486 | XP_016864368.1 | |
| AB014560.1 | 12 | 1531 | ||
| AF051311.1 | 10 | 1312 | AAC15705.1 | |
| AF053535.1 | 11 | 1507 | AAC95292.1 | |
| AF070615.1 | 8 | 1011 | ||
| AF145284.1 | 12 | 1606 | AAD51932.1 | |
| AK291772.1 | 11 | 1542 | ||
| AK291786.1 | 12 | 1546 | ||
| BC011731.2 | 11 | 1530 | AAH11731.1 |
More Information
Set Membership: |
|

Genomic Map