Assay Details
Target Gene Details
Entrez Gene ID: | 619279 |
Gene Name: | zinc finger protein 704 |
Gene Aliases: |
Gig1 |
Location: |
Chr.8:80628451-80884416 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF704 | NM_001033723.2 | 9 | 2011 | NP_001028895.1 |
| XM_005251279.4 | 9 | 2642 | XP_005251336.2 | |
| XM_017013725.1 | 9 | 2058 | XP_016869214.1 | |
| AK131274.1 | 9 | 2011 | BAD18451.1 | |
| BC136844.1 | 9 | 1824 | ||
| BC136845.1 | 9 | 1824 |
More Information
Set Membership: |
|

Genomic Map