Assay Details
Target Gene Details
Entrez Gene ID: | 5929 |
Gene Name: | RB binding protein 5, histone lysine methyltransferase complex subunit |
Gene Aliases: |
RBQ3, SWD1 |
Location: |
Chr.1:205086142-205122022 on Build GRCh38 |
Assay Gene Location: | Within Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RBBP5 | NM_001193272.1 | 14 | 2424 | NP_001180201.1 |
| NM_001193273.1 | 14 | 2479 | NP_001180202.1 | |
| NM_005057.3 | 14 | 2538 | NP_005048.2 | |
| XM_011509833.2 | 14 | 2533 | XP_011508135.1 | |
| BC037284.2 | 14 | 2377 | AAH37284.1 | |
| BC053856.1 | 14 | 2509 | AAH53856.1 | |
| X85134.1 | 14 | 2504 | CAA59446.1 |
More Information
Set Membership: |
|

Genomic Map