Assay Details
Target Gene Details
Entrez Gene ID: | 23630 |
Gene Name: | potassium voltage-gated channel subfamily E regulatory subunit 5 |
Gene Aliases: |
KCNE1L |
Location: |
Chr.X:109623700-109625164 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNE5 | NM_012282.2 | 1 | 1284 | NP_036414.1 |
| AF086499.1 | 1 | 136 | ||
| AJ012743.1 | 1 | 1284 | CAB58359.1 | |
| AK223306.1 | 1 | 1288 | BAD97026.1 | |
| BC035330.1 | 1 | 1263 | AAH35330.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv7033 | Chr.X:109601071 - 109632948 on Build GRCh38 | Insertion |
|
| esv2758883 | Chr.X:109535772 - 109737663 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map