Assay Details
Target Gene Details
Entrez Gene ID: | 55808 |
Gene Name: | ST6 N-acetylgalactosaminide alpha-2,6-sialyltransferase 1 |
Gene Aliases: |
HSY11339, SIAT7A, ST6GalNAcI, STYI |
Location: |
Chr.17:76617769-76643838 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ST6GALNAC1 | NM_001289107.1 | 3 | 622 | NP_001276036.1 |
| NM_018414.4 | 2 | 485 | NP_060884.1 | |
| NR_110309.1 | 2 | 485 | ||
| XM_011524995.2 | 2 | 405 | XP_011523297.1 | |
| XM_011524996.2 | 2 | 405 | XP_011523298.1 | |
| XM_017024842.1 | 2 | 405 | XP_016880331.1 | |
| XM_017024843.1 | 2 | 405 | XP_016880332.1 | |
| XM_017024844.1 | 3 | 505 | XP_016880333.1 | |
| AK310906.1 | 2 | 421 | ||
| AY096001.1 | 2 | 285 | AAM22800.1 | |
| AY358918.1 | 2 | 430 | AAQ89277.1 | |
| BC022462.1 | 2 | 340 | AAH22462.1 | |
| DA594489.1 | 2 | 485 | ||
| Y11339.2 | 2 | 345 | CAA72179.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3641295 | Chr.17:76628559 - 76633128 on Build GRCh38 | Gain |
|
| dgv588e214 | Chr.17:76627849 - 76633273 on Build GRCh38 | Loss |
|
| nsv1060182 | Chr.17:76597910 - 76660156 on Build GRCh38 | Gain |
|
| nsv518385 | Chr.17:76597821 - 76661078 on Build GRCh38 | Gain |
|
| esv2716257 | Chr.17:76627883 - 76633463 on Build GRCh38 | Deletion |
|
| esv2671702 | Chr.17:76627871 - 76633339 on Build GRCh38 | Deletion |
|
| nsv1058486 | Chr.17:76592348 - 76634542 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map