Assay Details
Target Gene Details
Entrez Gene ID: | 8703 |
Gene Name: | beta-1,4-galactosyltransferase 3 |
Gene Aliases: |
beta4Gal-T3 |
Location: |
Chr.1:161171310-161177968 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B4GALT3 | NM_001199873.1 | 8 | 2152 | NP_001186802.1 |
| NM_001199874.1 | 8 | 2081 | NP_001186803.1 | |
| NM_003779.3 | 8 | 2131 | NP_003770.1 | |
| XM_005245566.1 | 9 | 1853 | XP_005245623.1 | |
| XM_011510093.1 | 8 | 1662 | XP_011508395.1 | |
| XM_017002714.1 | 8 | 1683 | XP_016858203.1 | |
| AF038661.1 | 8 | 1658 | AAC39734.1 | |
| AK023311.1 | 9 | 1591 | BAB14520.1 | |
| AK056851.1 | 6 | 2819 | ||
| AK092493.1 | 9 | 1613 | ||
| BC000276.1 | 8 | 1634 | AAH00276.1 | |
| BC006099.1 | 8 | 1648 | AAH06099.1 | |
| BC009985.2 | 8 | 1610 | AAH09985.1 |
Target Gene Details
Entrez Gene ID: | 5498 |
Gene Name: | protoporphyrinogen oxidase |
Gene Aliases: |
PPO, V290M, VP |
Location: |
Chr.1:161165728-161178277 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
More Information
Set Membership: |
|

Genomic Map