Assay Details
Target Gene Details
Entrez Gene ID: | 890 |
Gene Name: | cyclin A2 |
Gene Aliases: |
CCN1, CCNA |
Location: |
Chr.4:121816444-121823933 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNA2 | NM_001237.3 | 5 | 1184 | NP_001228.1 |
| AK291931.1 | 5 | 1167 | ||
| BC104783.1 | 5 | 979 | AAI04784.1 | |
| BC104787.1 | 5 | 979 | AAI04788.1 | |
| CR407692.1 | 5 | 879 | CAG28620.1 | |
| X51688.1 | 5 | 986 | CAA35986.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv997539 | Chr.4:121622949 - 122353111 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map