Assay Details
Target Gene Details
Entrez Gene ID: | 59341 |
Gene Name: | transient receptor potential cation channel subfamily V member 4 |
Gene Aliases: |
BCYM3, CMT2C, HMSN2C, OTRPC4, SMAL, SPSMA, SSQTL1, TRP12, VRL2, VROAC |
Location: |
Chr.12:109783087-109833407 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TRPV4 | NM_001177428.1 | 14 | 2981 | NP_001170899.1 |
| NM_001177431.1 | 16 | 2989 | NP_001170902.1 | |
| NM_001177433.1 | 13 | 2801 | NP_001170904.1 | |
| NM_021625.4 | 16 | 3180 | NP_067638.3 | |
| NM_147204.2 | 14 | 2942 | NP_671737.1 | |
| XM_005253918.1 | 16 | 3540 | XP_005253975.1 | |
| XM_011538630.2 | 16 | 4283 | XP_011536932.2 | |
| XM_011538631.2 | 15 | 4143 | XP_011536933.2 | |
| XM_011538632.2 | 15 | 4104 | XP_011536934.2 | |
| XM_011538633.2 | 14 | 3963 | XP_011536935.2 | |
| XM_017019774.1 | 16 | 3913 | XP_016875263.1 | |
| AB032427.1 | 15 | 3186 | BAB69040.1 | |
| AJ296305.1 | 16 | 3180 | CAC82937.1 | |
| AL834192.1 | 16 | 3152 |
More Information
Set Membership: |
|

Genomic Map