Assay Details
Target Gene Details
Entrez Gene ID: | 156 |
Gene Name: | G protein-coupled receptor kinase 2 |
Gene Aliases: |
ADRBK1, BARK1, BETA-ARK1 |
Location: |
Chr.11:67266420-67286558 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GRK2 | NM_001619.3 | NP_001610.2 | ||
| XM_011544773.1 | XP_011543075.1 | |||
| AB209588.1 | 6 | 1502 | BAD92825.1 | |
| AK131506.1 | 1 | 915 | ||
| BC037963.1 | AAH37963.1 | |||
| BC090863.1 | AAH90863.1 | |||
| KU177896.1 | ||||
| KU177897.1 | ||||
| M80776.1 | AAA58391.1 | |||
| X61157.1 | CAA43470.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv555253 | Chr.11:67265605 - 67397024 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map