Assay Details
Target Gene Details
Entrez Gene ID: | 56204 |
Gene Name: | family with sequence similarity 214 member A |
Gene Aliases: |
KIAA1370 |
Location: |
Chr.15:52581321-52709817 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM214A | NM_001286495.1 | 5 | 2043 | NP_001273424.1 |
| NM_019600.3 | 6 | 2042 | NP_062546.2 | |
| NR_104457.1 | 6 | 2184 | ||
| XM_005254547.2 | 6 | 2005 | XP_005254604.1 | |
| XM_005254548.2 | 6 | 2524 | XP_005254605.1 | |
| XM_011521789.2 | 6 | 2071 | XP_011520091.1 | |
| XM_011521790.1 | 5 | 2043 | XP_011520092.1 | |
| AB037791.1 | 6 | 2020 | ||
| AK292917.1 | 6 | 2031 | ||
| AK293730.1 | 1 | 1314 | ||
| AK293920.1 | 5 | 2033 | ||
| AK308049.1 | 5 | 1848 | ||
| BC109127.1 | 5 | 1973 | AAI09128.1 | |
| BC109128.2 | 5 | 1973 | AAI09129.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833006 | Chr.15:52533757 - 52748265 on Build GRCh38 | Gain |
|
| nsv1047775 | Chr.15:52529853 - 52620031 on Build GRCh38 | Gain |
|
| nsv471245 | Chr.15:52315065 - 52894968 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs566843833] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map