Assay Details
Target Gene Details
Entrez Gene ID: | 92591 |
Gene Name: | ankyrin repeat and SOCS box containing 16 |
Gene Aliases: |
- |
Location: |
Chr.17:44170706-44179083 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASB16 | NM_080863.4 | 2 | 472 | NP_543139.4 |
| AF403034.1 | 1 | 87 | AAL57353.1 | |
| AK054727.1 | 2 | 472 | BAB70800.1 | |
| AK314595.1 | 2 | 464 | ||
| AY557346.1 | 2 | 388 | AAS67920.1 | |
| BC075088.2 | 2 | 421 | AAH75088.1 | |
| BC118584.1 | 2 | 454 | ||
| BC118585.1 | 2 | 454 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833455 | Chr.17:44108553 - 44297300 on Build GRCh38 | Loss |
|
| dgv529n67 | Chr.17:44170904 - 44246444 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map