Assay Details
Target Gene Details
Entrez Gene ID: | 5315 |
Gene Name: | pyruvate kinase, muscle |
Gene Aliases: |
CTHBP, HEL-S-30, OIP3, PK3, PKM2, TCB, THBP1 |
Location: |
Chr.15:72199029-72231624 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PKM | NM_001206796.2 | 1 | 422 | NP_001193725.1 |
| NM_001206797.2 | 1 | 422 | NP_001193726.1 | |
| NM_001206798.2 | 1 | 422 | NP_001193727.1 | |
| NM_001316318.1 | 1 | 422 | NP_001303247.1 | |
| NM_002654.5 | 1 | 422 | NP_002645.3 | |
| NM_182470.3 | 1 | 422 | NP_872270.1 | |
| NM_182471.3 | 1 | 422 | NP_872271.1 | |
| XM_005254445.4 | XP_005254502.1 | |||
| XM_011521670.2 | XP_011519972.1 | |||
| XM_017022313.1 | XP_016877802.1 | |||
| AK092369.1 | ||||
| AK222927.1 | BAD96647.1 | |||
| AK299413.1 | ||||
| AK300800.1 | ||||
| AK307855.1 | ||||
| AK312253.1 | ||||
| BC000481.2 | ||||
| BC007640.1 | AAH07640.1 | |||
| BC007952.2 | ||||
| BC012811.2 | ||||
| BC035198.1 | AAH35198.1 | |||
| BC094767.1 | AAH94767.1 | |||
| BG490968.1 | 1 | 180 | ||
| DA382029.1 | ||||
| DC326232.1 | ||||
| DC376054.1 | 1 | 184 | ||
| EU794631.1 | 1 | 141 | ||
| M23725.1 | AAA36449.1 | |||
| M26252.1 | AAA36672.1 | |||
| Z36831.1 |
More Information
Set Membership: |
|

Genomic Map