Assay Details
Target Gene Details
Entrez Gene ID: | 6830 |
Gene Name: | SPT6 homolog, histone chaperone |
Gene Aliases: |
SPT6, SPT6H, emb-5 |
Location: |
Chr.17:28662183-28702684 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SUPT6H | NM_001320755.1 | NP_001307684.1 | ||
| NM_003170.4 | NP_003161.2 | |||
| XM_017024978.1 | XP_016880467.1 | |||
| XM_017024979.1 | XP_016880468.1 | |||
| XM_017024980.1 | XP_016880469.1 | |||
| AL833607.1 | ||||
| BC003696.1 | AAH03696.1 | |||
| BC033074.1 | AAH33074.1 | |||
| BC136522.1 | ||||
| BC136524.1 | ||||
| BC150268.1 | ||||
| D79984.1 | ||||
| U38623.1 | AAC99996.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map