Assay Details
Target Gene Details
Entrez Gene ID: | 10557 |
Gene Name: | ribonuclease P/MRP subunit p38 |
Gene Aliases: |
- |
Location: |
Chr.10:15097180-15104257 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RPP38 | NM_001097590.2 | 3 | 938 | NP_001091059.1 |
| NM_001265601.1 | 2 | 1176 | NP_001252530.1 | |
| NM_006414.4 | 2 | 819 | NP_006405.2 | |
| NM_183005.4 | 3 | 1295 | NP_892117.1 | |
| XM_006717363.1 | 3 | 1109 | XP_006717426.1 | |
| XM_006717364.3 | 2 | 731 | XP_006717427.1 | |
| XM_011519293.1 | 2 | 974 | XP_011517595.1 | |
| XM_017015479.1 | 3 | 1427 | XP_016870968.1 | |
| XM_017015480.1 | 3 | 885 | XP_016870969.1 | |
| XM_017015481.1 | 2 | 909 | XP_016870970.1 | |
| AK057006.1 | 3 | 1093 | ||
| AK223418.1 | 2 | 796 | BAD97138.1 | |
| BC029494.1 | 3 | 1076 | AAH29494.1 | |
| CR541816.1 | 1 | 579 | CAG46615.1 | |
| U77664.1 | 2 | 615 | AAC51142.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831794 | Chr.10:14993658 - 15148891 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map