Assay Details
Target Gene Details
Entrez Gene ID: | 51703 |
Gene Name: | acyl-CoA synthetase long-chain family member 5 |
Gene Aliases: |
ACS2, ACS5, FACL5 |
Location: |
Chr.10:112374158-112428380 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 21 - Intron 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACSL5 | NM_016234.3 | NP_057318.2 | ||
| NM_203379.1 | NP_976313.1 | |||
| NM_203380.1 | NP_976314.1 | |||
| XM_017016324.1 | XP_016871813.1 | |||
| AB033899.1 | BAA85979.1 | |||
| AK000339.1 | ||||
| AK024932.1 | ||||
| AK222782.1 | ||||
| AK223288.1 | ||||
| AK301786.1 | ||||
| AK310436.1 | ||||
| AM262166.1 | ||||
| AY358520.1 | AAQ88884.1 | |||
| BC007985.2 | AAH07985.2 |
Target Gene Details
Entrez Gene ID: | 64429 |
Gene Name: | zinc finger DHHC-type containing 6 |
Gene Aliases: |
DHHC-6, ZNF376 |
Location: |
Chr.10:112424411-112448335 on Build GRCh38 |
Assay Gene Location: | Within Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZDHHC6 | XM_011540091.2 | XP_011538393.1 | ||
| XM_017016564.1 | XP_016872053.1 | |||
| XM_017016565.1 | XP_016872054.1 | |||
| XM_017016566.1 | XP_016872055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831990 | Chr.10:112349267 - 112531997 on Build GRCh38 | Loss |
|
| nsv831991 | Chr.10:112413098 - 112582139 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map