Assay Details
Target Gene Details
Entrez Gene ID: | 55716 |
Gene Name: | limb development membrane protein 1 like |
Gene Aliases: |
LIMR |
Location: |
Chr.12:49096551-49111535 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629447 | Chr.12:49099548 - 49107982 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map