Assay Details
Target Gene Details
Entrez Gene ID: | 5162 |
Gene Name: | pyruvate dehydrogenase (lipoamide) beta |
Gene Aliases: |
PDHBD, PDHE1-B, PHE1B |
Location: |
Chr.3:58427630-58433852 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDHB | NM_000925.3 | 7 | 698 | NP_000916.2 |
| NM_001173468.1 | 8 | 644 | NP_001166939.1 | |
| NM_001315536.1 | 6 | 644 | NP_001302465.1 | |
| NR_033384.1 | 6 | 768 | ||
| AB209241.1 | 4 | 3060 | BAD92478.1 | |
| AK225569.1 | 7 | 673 | ||
| AK225621.1 | 7 | 678 | ||
| AK293153.1 | 8 | 610 | ||
| AK311514.1 | 8 | 549 | ||
| AK313022.1 | 7 | 673 | ||
| AL117618.1 | 6 | 609 | CAB56017.1 | |
| BC000439.2 | 7 | 665 | AAH00439.1 | |
| BC001924.1 | 7 | 665 | AAH01924.1 | |
| CR541911.1 | 7 | 655 | CAG46709.1 | |
| J03576.1 | 7 | 673 | AAA88097.1 | |
| M34055.1 | 7 | 660 | AAA60233.1 | |
| M34056.1 | 6 | 583 | AAA60054.1 | |
| M34479.1 | 7 | 655 | AAA36428.1 | |
| M54788.1 | 7 | 660 | AAA60053.1 | |
| X57778.1 | 7 | 674 | CAA40924.1 |
More Information
Set Membership: |
|

Genomic Map