Assay Details
Target Gene Details
Entrez Gene ID: | 51388 |
Gene Name: | NIP7, nucleolar pre-rRNA processing protein |
Gene Aliases: |
CGI-37, HSPC031, KD93 |
Location: |
Chr.16:69339512-69343111 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NIP7 | NM_001199434.1 | NP_001186363.1 | ||
| NM_016101.4 | NP_057185.1 | |||
| AB112439.2 | BAD05056.1 | |||
| AF161528.1 | AAF29143.1 | |||
| AW001672.1 |
More Information
Set Membership: |
|

Genomic Map