Assay Details
Target Gene Details
Entrez Gene ID: | 7469 |
Gene Name: | negative elongation factor complex member A |
Gene Aliases: |
NELF-A, P/OKcl.15, WHSC2 |
Location: |
Chr.4:1982714-2009972 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NELFA | NM_005663.4 | 11 | 1995 | NP_005654.3 |
| XM_017008589.1 | 12 | 2816 | XP_016864078.1 | |
| AB044549.1 | 10 | 1782 | BAB18651.1 | |
| AF101434.1 | 11 | 1724 | AAC72982.1 | |
| AF131751.1 | 7 | 1245 | AAD20034.1 | |
| AK001304.1 | 11 | 1782 | ||
| AK094040.1 | 10 | 1555 | ||
| AK095737.1 | 10 | 2038 | ||
| AK126056.1 | 11 | 2870 | ||
| AK222853.1 | 11 | 1732 | BAD96573.1 | |
| BC002764.2 | 11 | 1735 | AAH02764.2 |
More Information
Set Membership: |
|

Genomic Map