Assay Details
Target Gene Details
Entrez Gene ID: | 10393 |
Gene Name: | anaphase promoting complex subunit 10 |
Gene Aliases: |
APC10, DOC1 |
Location: |
Chr.4:144967117-145098541 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
More Information
Set Membership: |
|

Genomic Map