Assay Details
Target Gene Details
Entrez Gene ID: | 2161 |
Gene Name: | coagulation factor XII |
Gene Aliases: |
HAE3, HAEX, HAF |
Location: |
Chr.5:177402138-177409576 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| F12 | NM_000505.3 | NP_000496.2 | ||
| XM_011534462.2 | XP_011532764.1 | |||
| AB095845.1 | BAC23095.1 | |||
| BC012390.1 | AAH12390.1 | |||
| M11723.1 | AAA51986.1 | |||
| M13147.1 | AAA70224.1 | |||
| M31315.1 | AAA70225.1 |
Target Gene Details
Entrez Gene ID: | 6569 |
Gene Name: | solute carrier family 34 member 1 |
Gene Aliases: |
FRTS2, HCINF2, NAPI-3, NPHLOP1, NPT2, NPTIIa, SLC11, SLC17A2 |
Location: |
Chr.5:177384431-177412021 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC34A1 | XM_017009773.1 | XP_016865262.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv471057 | Chr.5:177310308 - 177469411 on Build GRCh38 | Loss |
|
| nsv509103 | Chr.5:177308751 - 177414575 on Build GRCh38 | Insertion |
|
| nsv600364 | Chr.5:177299634 - 177432847 on Build GRCh38 | Loss |
|
| dgv10185n54 | Chr.5:177398001 - 177406217 on Build GRCh38 | Loss |
|
| dgv10184n54 | Chr.5:177357511 - 177469411 on Build GRCh38 | Loss |
|
| nsv1161313 | Chr.5:177356552 - 177463415 on Build GRCh38 | Duplication |
|
| nsv1161312 | Chr.5:177322159 - 177405557 on Build GRCh38 | Deletion |
|
| nsv1161311 | Chr.5:177305104 - 177546474 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map