Assay Details
Target Gene Details
Entrez Gene ID: | 6764 |
Gene Name: | suppression of tumorigenicity 5 |
Gene Aliases: |
DENND2B, HTS1, p126 |
Location: |
Chr.11:8693352-8910951 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 27 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951306 | Chr.11:8701054 - 8735653 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map