Assay Details
Target Gene Details
Entrez Gene ID: | 93377 |
Gene Name: | oligodendrocytic myelin paranodal and inner loop protein |
Gene Aliases: |
HTMP10, TMEM10, TMP10 |
Location: |
Chr.10:96343216-96359365 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OPALIN | NM_001040102.2 | 4 | 3317 | NP_001035191.1 |
| NM_001040103.2 | 7 | 3425 | NP_001035192.1 | |
| NM_001284320.1 | 5 | 3353 | NP_001271249.1 | |
| NM_001284321.1 | 5 | 3350 | NP_001271250.1 | |
| NM_001284322.1 | 7 | 3468 | NP_001271251.1 | |
| NM_001284323.1 | 7 | 3203 | NP_001271252.1 | |
| NM_001284324.1 | 7 | 3096 | NP_001271253.1 | |
| NM_001284326.1 | 6 | 3164 | NP_001271255.1 | |
| NM_001284327.1 | 6 | 3057 | NP_001271256.1 | |
| NM_033207.4 | 6 | 3386 | NP_149984.1 | |
| NR_104296.1 | 6 | 3445 | ||
| NR_104297.1 | 6 | 3372 | ||
| AF367761.2 | 6 | 3326 | AAK62349.1 | |
| AL834229.1 | 6 | 3003 | CAD38907.1 | |
| BC033737.1 | 6 | 3003 | AAH33737.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831950 | Chr.10:96321215 - 96485677 on Build GRCh38 | Gain |
|
| nsv831949 | Chr.10:96167838 - 96385945 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs113594239] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map