Assay Details
Target Gene Details
Entrez Gene ID: | 23230 |
Gene Name: | vacuolar protein sorting 13 homolog A |
Gene Aliases: |
CHAC, CHOREIN |
Location: |
Chr.9:77177353-77421537 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS13A | NM_001018037.1 | 1 | 268 | NP_001018047.1 |
| NM_001018038.2 | 1 | 268 | NP_001018048.1 | |
| NM_015186.3 | 1 | 268 | NP_056001.1 | |
| NM_033305.2 | 1 | 268 | NP_150648.2 | |
| AB054005.1 | 1 | 179 | BAB59128.1 | |
| AF337532.1 | 1 | 268 | AAK61861.1 | |
| AJ608769.1 | 1 | 268 | CAE75581.1 | |
| AJ626859.1 | 1 | 268 | CAF25186.1 | |
| AW303378.1 | 1 | 262 | ||
| BI828209.1 | 1 | 444 |
Target Gene Details
Entrez Gene ID: | 100286938 |
Gene Name: | VPS13A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:77176756-77177917 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VPS13A-AS1 | NR_026668.2 | 1 | 206 | |
| AA626167.1 | 2 | 233 | ||
| DB509755.1 | 1 | 212 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7682n100 | Chr.9:77154478 - 77180761 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map