Assay Details
Target Gene Details
Entrez Gene ID: | 26002 |
Gene Name: | monooxygenase DBH like 1 |
Gene Aliases: |
MOX, PRO5780, dJ248E1.1 |
Location: |
Chr.6:132296055-132401534 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MOXD1 | NM_015529.3 | NP_056344.2 | ||
| XM_017010714.1 | XP_016866203.1 | |||
| XM_017010715.1 | XP_016866204.1 | |||
| AK074879.1 | BAC11263.1 | |||
| AK226054.1 | ||||
| AL080058.1 | CAB45692.1 | |||
| AY007239.1 | AAG09636.1 | |||
| AY359094.1 | AAQ89452.1 | |||
| BC018756.1 | AAH18756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830811 | Chr.6:132302021 - 132466400 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map