Assay Details
Target Gene Details
Entrez Gene ID: | 146894 |
Gene Name: | CD300 molecule like family member g |
Gene Aliases: |
CLM-9, CLM9, NEPMUCIN, TREM-4, TREM4 |
Location: |
Chr.17:43846777-43863639 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD300LG | NM_001168322.1 | 3 | 496 | NP_001161794.1 |
| NM_001168323.1 | NP_001161795.1 | |||
| NM_001168324.1 | NP_001161796.1 | |||
| NM_145273.3 | 3 | 496 | NP_660316.2 | |
| XM_005257075.2 | 3 | 867 | XP_005257132.1 | |
| XM_005257076.2 | XP_005257133.1 | |||
| XM_006721721.2 | 3 | 867 | XP_006721784.1 | |
| XM_006721722.2 | XP_006721785.1 | |||
| XM_011524384.2 | 3 | 867 | XP_011522686.1 | |
| XM_017024235.1 | XP_016879724.1 | |||
| AF427619.1 | AAN86134.1 | |||
| AF427620.1 | AAN86135.1 | |||
| AK298112.1 | ||||
| AK309938.1 | 3 | 497 | ||
| AY358364.1 | 3 | 451 | AAQ88730.1 | |
| BC025395.2 | 3 | 468 | AAH25395.1 |
More Information
Set Membership: |
|

Genomic Map