Assay Details
Target Gene Details
Entrez Gene ID: | 100507291 |
Gene Name: | uncharacterized LOC100507291 |
Gene Aliases: |
- |
Location: |
Chr.3:139389803-139583319 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100507291 | NR_121609.1 | |||
| DA364121.1 | ||||
| DA756291.1 |
Target Gene Details
Entrez Gene ID: | 349565 |
Gene Name: | nicotinamide nucleotide adenylyltransferase 3 |
Gene Aliases: |
FKSG76, PNAT-3, PNAT3 |
Location: |
Chr.3:139560180-139678050 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
More Information
Set Membership: |
|

Genomic Map