Assay Details
Target Gene Details
Entrez Gene ID: | 2277 |
Gene Name: | c-fos induced growth factor |
Gene Aliases: |
VEGF-D, VEGFD |
Location: |
Chr.X:15345591-15384413 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FIGF | NM_004469.4 | 7 | 1667 | NP_004460.1 |
| AJ000185.1 | 7 | 1610 | CAA03942.1 | |
| AK223160.1 | 7 | 1660 | BAD96880.1 | |
| BC027948.1 | 7 | 1636 | AAH27948.1 | |
| D89630.1 | 7 | 1602 | BAA24264.1 | |
| Y12863.1 | 7 | 1443 | CAA73370.1 |
Target Gene Details
Entrez Gene ID: | 100532742 |
Gene Name: | PIR-FIGF readthrough |
Gene Aliases: |
- |
Location: |
Chr.X:15345591-15491309 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIR-FIGF | NR_037859.2 | 15 | 2175 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv6813 | Chr.X:15339080 - 15383961 on Build GRCh38 | Deletion |
|
| nsv528343 | Chr.X:15271931 - 15385229 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map