Assay Details
Target Gene Details
Entrez Gene ID: | 117159 |
Gene Name: | dermcidin |
Gene Aliases: |
AIDD, DCD-1, DSEP, HCAP, PIF |
Location: |
Chr.12:54644591-54648493 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DCD | NM_001300854.1 | 1 | 186 | NP_001287783.1 |
| NM_053283.3 | 1 | 186 | NP_444513.1 | |
| AF144011.1 | AAL18349.1 | |||
| AY590150.1 | AAS99907.1 | |||
| BC062682.1 | AAH62682.1 | |||
| BC069108.1 | AAH69108.1 | |||
| BP313072.1 | ||||
| EF503687.1 | 1 | 186 | ||
| EF503688.1 | 1 | 186 | ||
| EF503689.1 | 1 | 186 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832421 | Chr.12:54522913 - 54691618 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map