Assay Details
Target Gene Details
Entrez Gene ID: | 10121 |
Gene Name: | ARP1 actin-related protein 1 homolog A, centractin alpha |
Gene Aliases: |
ARP1, Arp1A, CTRN1 |
Location: |
Chr.10:102479229-102502755 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACTR1A | NM_005736.3 | NP_005727.1 | ||
| AK297362.1 | ||||
| AK297620.1 | ||||
| AK312506.1 | ||||
| DC384284.1 | 1 | 88 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831968 | Chr.10:102482950 - 102664645 on Build GRCh38 | Gain |
|
| nsv467445 | Chr.10:102501602 - 102627177 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map