Assay Details
Target Gene Details
Entrez Gene ID: | 645 |
Gene Name: | biliverdin reductase B |
Gene Aliases: |
BVRB, FLR, HEL-S-10, SDR43U1 |
Location: |
Chr.19:40447784-40465818 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BLVRB | NM_000713.2 | NP_000704.1 | ||
| AK312137.1 | ||||
| BC109371.1 | AAI09372.1 | |||
| BG340491.1 | ||||
| D26308.1 | BAA05370.1 | |||
| D32143.1 | BAA06874.1 | |||
| EU794593.1 |
More Information
Set Membership: |
|

Genomic Map