Assay Details
Target Gene Details
Entrez Gene ID: | 284129 |
Gene Name: | solute carrier family 26 member 11 |
Gene Aliases: |
- |
Location: |
Chr.17:80220287-80253509 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC26A11 | NM_001166347.1 | 4 | 669 | NP_001159819.1 |
| NM_001166348.1 | 3 | 599 | NP_001159820.1 | |
| NM_001166349.1 | 3 | 469 | NP_001159821.1 | |
| NM_173626.3 | 4 | 666 | NP_775897.3 | |
| XM_006721833.1 | 4 | 669 | XP_006721896.1 | |
| XM_011524652.1 | 3 | 573 | XP_011522954.1 | |
| XM_011524653.1 | 3 | 573 | XP_011522955.1 | |
| XM_011524654.1 | 3 | 458 | XP_011522956.1 | |
| XM_017024505.1 | 3 | 583 | XP_016879994.1 | |
| XM_017024506.1 | 3 | 583 | XP_016879995.1 | |
| XM_017024507.1 | 4 | 643 | XP_016879996.1 | |
| AF345195.1 | 3 | 401 | AAO26673.1 | |
| AJ544073.1 | 4 | 640 | CAD66450.1 | |
| AK075248.1 | 4 | 629 | BAC11496.1 | |
| AK315132.1 | 4 | 632 | ||
| AL833468.1 | 3 | 563 | ||
| BC035900.1 | 4 | 608 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1066606 | Chr.17:79955666 - 80224006 on Build GRCh38 | Gain |
|
| nsv469591 | Chr.17:80085322 - 80293486 on Build GRCh38 | Loss |
|
| nsv828123 | Chr.17:80222547 - 80257700 on Build GRCh38 | Gain |
|
| nsv457968 | Chr.17:80205094 - 80292960 on Build GRCh38 | Loss |
|
| nsv833562 | Chr.17:80114523 - 80301339 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map