Assay Details
Target Gene Details
Entrez Gene ID: | 5576 |
Gene Name: | protein kinase cAMP-dependent type II regulatory subunit alpha |
Gene Aliases: |
PKR2, PRKAR2 |
Location: |
Chr.3:48744601-48847850 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRKAR2A | NM_001321982.1 | NP_001308911.1 | ||
| NM_001321983.1 | NP_001308912.1 | |||
| NM_001321989.1 | NP_001308918.1 | |||
| NM_004157.3 | NP_004148.1 | |||
| XM_005265315.3 | XP_005265372.1 | |||
| XM_011533942.2 | XP_011532244.1 | |||
| XM_011533943.1 | XP_011532245.1 | |||
| XM_017006815.1 | XP_016862304.1 | |||
| XM_017006816.1 | XP_016862305.1 | |||
| AK293042.1 | ||||
| BC002763.2 | AAH02763.1 | |||
| BT007225.1 | AAP35889.1 | |||
| KU178276.1 | ||||
| KU178277.1 | ||||
| X14968.1 | CAA33094.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508918 | Chr.3:48696212 - 48784507 on Build GRCh38 | Insertion |
|
| nsv590247 | Chr.3:48624552 - 48764496 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map