Assay Details
Target Gene Details
Entrez Gene ID: | 10053 |
Gene Name: | adaptor related protein complex 1 mu 2 subunit |
Gene Aliases: |
AP1-mu2, HSMU1B, MU-1B, MU1B, mu2 |
Location: |
Chr.19:10572671-10587315 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AP1M2 | NM_001300887.1 | 2 | 206 | NP_001287816.1 |
| NM_005498.4 | 2 | 206 | NP_005489.2 | |
| AF020797.2 | 2 | 169 | AAD25870.2 | |
| AK023863.1 | 2 | 207 | ||
| AK222946.1 | 2 | 172 | BAD96666.1 | |
| AK315689.1 | 2 | 197 | ||
| BC003387.2 | 2 | 144 | AAH03387.1 | |
| BC003612.2 | 2 | 143 | AAH03612.1 | |
| BC005021.1 | 2 | 206 | AAH05021.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833744 | Chr.19:10458964 - 10635033 on Build GRCh38 | Loss |
|
| nsv833745 | Chr.19:10539132 - 10727920 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map