Assay Details
Target Gene Details
Entrez Gene ID: | 54956 |
Gene Name: | poly(ADP-ribose) polymerase family member 16 |
Gene Aliases: |
ARTD15, C15orf30, pART15 |
Location: |
Chr.15:65231691-65286886 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PARP16 | NM_001316943.1 | NP_001303872.1 | ||
| NM_001316944.1 | NP_001303873.1 | |||
| NM_017851.5 | NP_060321.3 | |||
| XM_006720589.2 | XP_006720652.1 | |||
| XM_006720590.2 | XP_006720653.1 | |||
| XM_006720592.2 | XP_006720655.1 | |||
| XM_011521742.2 | XP_011520044.1 | |||
| XM_017022386.1 | XP_016877875.1 | |||
| XM_017022387.1 | XP_016877876.1 | |||
| XM_017022388.1 | XP_016877877.1 | |||
| XM_017022389.1 | XP_016877878.1 | |||
| AK000516.1 | BAA91222.1 | |||
| AK058010.1 | ||||
| BC006389.1 | AAH06389.1 | |||
| BC031074.1 | AAH31074.1 |
More Information
Set Membership: |
|

Genomic Map