Assay Details
Target Gene Details
Entrez Gene ID: | 255725 |
Gene Name: | olfactory receptor family 52 subfamily B member 2 |
Gene Aliases: |
OR11-70 |
Location: |
Chr.11:6169355-6170326 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR52B2 | NM_001004052.1 | 1 | 678 | NP_001004052.1 |
| BC136732.1 | 1 | 760 | ||
| BC136733.1 | 1 | 760 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv28312 | Chr.11:6168480 - 6171866 on Build GRCh38 | Loss |
|
| esv3625262 | Chr.11:6168439 - 6171798 on Build GRCh38 | Loss |
|
| nsv523933 | Chr.11:6152116 - 6183086 on Build GRCh38 | Loss |
|
| esv2658298 | Chr.11:6168354 - 6171844 on Build GRCh38 | Deletion |
|
| esv2421684 | Chr.11:6168573 - 6171232 on Build GRCh38 | Deletion |
|
| esv3891923 | Chr.11:6154881 - 6183086 on Build GRCh38 | Loss |
|
| nsv553241 | Chr.11:5484804 - 6182455 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77344022] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map