Assay Details
Target Gene Details
Entrez Gene ID: | 731 |
Gene Name: | complement component 8 alpha subunit |
Gene Aliases: |
- |
Location: |
Chr.1:56854770-56918221 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C8A | NM_000562.2 | 5 | 712 | NP_000553.1 |
| XM_011542079.2 | 5 | 686 | XP_011540381.1 | |
| XM_017002234.1 | 5 | 686 | XP_016857723.1 | |
| AK309596.1 | 5 | 686 | ||
| BC020702.1 | 5 | 685 | ||
| BC132911.1 | 5 | 604 | ||
| BC132913.1 | 5 | 604 | ||
| M16974.1 | 5 | 717 | AAA52200.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508215 | Chr.1:56853787 - 56885477 on Build GRCh38 | Deletion |
|
| esv3586132 | Chr.1:56880453 - 56883382 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs116129675] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map