Assay Details
Target Gene Details
Entrez Gene ID: | 3593 |
Gene Name: | interleukin 12B |
Gene Aliases: |
CLMF, CLMF2, IL-12B, IMD28, IMD29, NKSF, NKSF2 |
Location: |
Chr.5:159314783-159330473 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IL12B | NM_002187.2 | 3 | 250 | NP_002178.2 |
| AF180563.1 | 2 | 214 | AAD56386.1 | |
| BC067498.1 | 2 | 212 | AAH67498.1 | |
| BC067499.1 | 2 | 212 | AAH67499.1 | |
| BC067500.1 | 2 | 212 | AAH67500.1 | |
| BC067501.1 | 2 | 212 | AAH67501.1 | |
| BC067502.1 | 2 | 212 | AAH67502.1 | |
| BC074723.2 | 3 | 239 | AAH74723.1 | |
| EF173865.1 | ||||
| M65272.1 | 3 | 250 | AAA35695.1 | |
| M65290.1 | 2 | 221 | AAA59938.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3607337 | Chr.5:159276823 - 159478320 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map