Assay Details
Target Gene Details
Entrez Gene ID: | 390323 |
Gene Name: | olfactory receptor family 6 subfamily C member 75 |
Gene Aliases: |
- |
Location: |
Chr.12:55365111-55366049 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6C75 | NM_001005497.1 | 1 | 680 | NP_001005497.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1045950 | Chr.12:55360325 - 55440504 on Build GRCh38 | Gain |
|
| nsv559001 | Chr.12:55113681 - 55663924 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map