Assay Details
Target Gene Details
Entrez Gene ID: | 23538 |
Gene Name: | olfactory receptor family 52 subfamily A member 1 |
Gene Aliases: |
HPFH1OR |
Location: |
Chr.11:5151431-5152369 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR52A1 | NM_012375.2 | 1 | 426 | NP_036507.2 |
| BC111722.1 | 1 | 426 | AAI11723.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760169 | Chr.11:4940288 - 5212591 on Build GRCh38 | Gain+Loss |
|
| esv3625208 | Chr.11:5144423 - 5175233 on Build GRCh38 | Loss |
|
| nsv553219 | Chr.11:5004970 - 5193183 on Build GRCh38 | Gain |
|
| esv3891917 | Chr.11:5081246 - 5227774 on Build GRCh38 | Loss |
|
| nsv553224 | Chr.11:5117503 - 5186159 on Build GRCh38 | Loss |
|
| esv3625206 | Chr.11:5114340 - 5159347 on Build GRCh38 | Loss |
|
| nsv1054847 | Chr.11:4621645 - 5179426 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77308879] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map