Assay Details
Target Gene Details
Entrez Gene ID: | 55930 |
Gene Name: | myosin VC |
Gene Aliases: |
- |
Location: |
Chr.15:52192318-52295798 on Build GRCh38 |
Assay Gene Location: | Within Exon 32 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO5C | NM_018728.3 | 32 | 4018 | NP_061198.2 |
| XM_011521781.2 | 32 | 3934 | XP_011520083.1 | |
| XM_017022408.1 | 31 | 3883 | XP_016877897.1 | |
| XM_017022409.1 | 27 | 3325 | XP_016877898.1 | |
| XM_017022410.1 | 31 | 3868 | XP_016877899.1 | |
| AF272390.1 | 32 | 3875 | AAF78783.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1537 | Chr.15:52210049 - 52244870 on Build GRCh38 | Deletion |
|
| nsv471244 | Chr.15:52013266 - 52219268 on Build GRCh38 | Gain |
|
| nsv833005 | Chr.15:52097672 - 52249133 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map